Assay Details
| Gene Symbol: PRKCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130736.1 | - | 1 | 1851 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PRKCA antisense RNA 1 | PRKCA-AS1 | -279.5kb | Anti-sense | Chr. 17 - 66398069 - 66416854 on NCBI Build GRCh38 | |
| Downstream | microRNA 634 | MIR634 |
MIRN634, hsa-mir-634 |
88.3kb | Sense | Chr. 17 - 66787072 - 66787168 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PRKCA
Genomic Map