Assay Details
| Gene Symbol: ARHGAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130998.1 | - | 1 | 51 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4799 | MIR4799 | -127.1kb | Sense | Chr. 4 - 147782595 - 147782668 on NCBI Build GRCh38 | |
| Downstream | nuclear receptor subfamily 3 group C member 2 | NR3C2 |
MCR, MLR, MR, NR3C2VIT |
168.4kb | Anti-sense | Chr. 4 - 148078764 - 148445287 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ARHGAP10
Genomic Map