Assay Details
| Gene Symbol: EEF1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123690.1 | - | 1 | 2653 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | maestro heat like repeat family member 8 | MROH8 |
C20orf131, C20orf132, dJ621N11.3, dJ621N11.4 |
-58.6kb | Anti-sense | Chr. 20 - 37101226 - 37179588 on NCBI Build GRCh38 |
| Downstream | growth hormone releasing hormone | GHRH |
GHRF, GRF, INN |
9.7kb | Anti-sense | Chr. 20 - 37251086 - 37261817 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

EEF1A2
Genomic Map