Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012088.hg.1 |
ENST00000421194.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012088.hg.1 |
ENST00000421194.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012088.hg.1 |
ENST00000626603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012088.hg.1 |
ENST00000626603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012088.hg.1 |
ENST00000626603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012088.hg.1 |
ENST00000629344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012088.hg.1 |
ENST00000629344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012088.hg.1 |
ENST00000421194.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012088.hg.1 |
ENST00000421194.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012088.hg.1 |
ENST00000626603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012088.hg.1 |
ENST00000626603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012088.hg.1 |
ENST00000626603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012088.hg.1 |
ENST00000629344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012088.hg.1 |
ENST00000629344.1 |
Ensembl |
Human Genome U95 Set |
90081_at |
- |
- |
Human Genome U95 Set |
90081_PM_at |
- |
- |