Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008389.hg.1 |
ENST00000611513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008389.hg.1 |
ENST00000611513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008389.hg.1 |
ENST00000611513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000547140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000547140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000547140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000548755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000548755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
ENST00000548755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008389.hg.1 |
ENST00000611513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008389.hg.1 |
ENST00000611513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008389.hg.1 |
ENST00000611513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000279488.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000308385.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000547140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000547140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000547140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000547291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000548755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000548755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
ENST00000548755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_001946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011470.hg.1 |
NM_022652.3 |
Refseq |
Human Genome U95 Set |
48316_PM_at |
- |
- |
Human Genome U95 Set |
48316_at |
- |
- |