Assay Details
| Gene Symbol: LOC100131242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124483.1 | - | 1 | 2055 | 58 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 38 member 10 | SLC38A10 | PP1744 | Anti-sense | Chr. 17 - 81244811 - 81295433 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 482 | LINC00482 | C17orf55 | 37.4kb | Anti-sense | Chr. 17 - 81302824 - 81309248 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100131242
Genomic Map