Assay Details
| Gene Symbol: LOC100128401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124497.1 | - | 1 | 1856 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2 |
HPS4 |
BLOC3S2, LE |
Sense | Chr. 22 - 26443613 - 26483863 on NCBI Build GRCh38 | |
| Downstream | SRR1 domain containing | SRRD |
HC/HCC, SRR1L |
37.0kb | Anti-sense | Chr. 22 - 26483867 - 26494620 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100128401
Genomic Map