Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000471328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000471328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000477004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000477004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000477004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000477004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000512382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000512382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000512382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000512382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000313766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000471328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000471328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000477004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000477004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000477004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000477004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000512382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000512382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000512382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000512382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006445.hg.1 |
ENST00000515795.1 |
Ensembl |
Human Genome U133 Plus 2.0 Array |
229438_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
229438_PM_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229438_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229438_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229438_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229438_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229438_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229438_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229438_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229438_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229438_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229438_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229438_at10 |
- |
- |
Human Genome U95 Set |
67069_at |
- |
- |
Human Genome U95 Set |
67069_PM_at |
- |
- |
Human Genome U133 Set |
229438_at |
- |
- |
Human Genome U133 Set |
229438_PM_at |
- |
- |