Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027728.hg.1 |
TC03001524.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027728.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027728.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011385.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011385.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011385.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011385.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011385.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011385.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011385.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011385.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027729.hg.1 |
TC03001524.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027729.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027729.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027727.hg.1 |
TC03001524.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027727.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027727.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027730.hg.1 |
TC03001524.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027730.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027730.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185389.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185389.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185389.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185389.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185389.hg.1 |
TC0300011385.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011385.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011385.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011385.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011385.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011385.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011385.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011385.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011385.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185386.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185386.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185386.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185386.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185386.hg.1 |
TC0300011385.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300185388.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185388.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185388.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185388.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185388.hg.1 |
TC0300011385.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300185387.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185387.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185387.hg.1 |
ENST00000485805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185387.hg.1 |
NR_033978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185387.hg.1 |
TC0300011385.hg.1 |
Clariom D TC |