Assay Details
Gene Symbol: LINC01443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104164.1 | 3 - 4 | 462 | 87 | ||
GenBank mRNA | AK126243.1 | - | 3 - 4 | 462 | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1444 | LINC01444 | Anti-sense | Chr. 18 - 14969607 - 14970468 on NCBI Build GRCh38 | ||
Downstream | ankyrin repeat domain 30B pseudogene | LOC644669 | 339.8kb | Anti-sense | Chr. 18 - 15313556 - 15325919 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10560737 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10560747 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31157518 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10560760 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10560758 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10560776 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31157572 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31157554 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10560752 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10560729 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31157431 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10560779 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10560745 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31157539 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31157606 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31157482 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17046185 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10560780 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31157484 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10560767 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10560750 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31157463 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31157573 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31157598 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10560762 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31157428 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31157494 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31157453 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10560768 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 166620 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 12705 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 373344 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 158718 | AB085843.1 | genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 158718 | AB085845.1 | genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 158718 | AF016381.1 | genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 158718 | AY212933.1 | genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 158718 | AY382151.1 | genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 158718 | M15716.1 | genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 158718 | NM_000926.4 | refseq_mrna_human_NM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 158718 | X51730.1 | genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 497047 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 398658 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 323562 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 389329 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 54809 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 65667 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 631283 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 892657 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 83883 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 348401 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 69147 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 132496 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 44151 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 48844 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 377157 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 185039 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 405627 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 411799 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 221621 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 129944 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 17029 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 131750 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 142651 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 402413 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 499 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 19386 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 274783 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3565342 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 2783 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P12799116 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P12796257 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P12799116 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3565342 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560733 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560764 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157545 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560743 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560767 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046089 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157534 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157606 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560727 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560778 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157463 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157497 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157609 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157518 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560765 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157422 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157470 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157457 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157431 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157539 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560779 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157507 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157484 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157434 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560750 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560732 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046068 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157524 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157445 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157448 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157495 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560748 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157480 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157598 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560749 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560734 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157428 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157572 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046084 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046204 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560760 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560737 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046185 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157488 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560768 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560752 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157573 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560742 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560780 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157554 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157494 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157454 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560769 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157597 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560753 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157516 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157596 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560773 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046104 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157482 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157590 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157570 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560730 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560776 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157550 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157483 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560747 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560729 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046107 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560758 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157453 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046058 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046139 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560745 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046047 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157578 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560777 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046190 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046118 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560738 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560762 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157546 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157575 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560740 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560775 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560743 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157572 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046139 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157507 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560773 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157454 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157480 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046118 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560768 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560780 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157539 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560733 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157554 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560749 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560732 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560729 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560742 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157570 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157609 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560738 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157575 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157482 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560745 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560769 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560779 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560778 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157488 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157495 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157497 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560775 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157596 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157422 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157546 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560747 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157457 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157484 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560758 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046190 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157518 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157453 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560730 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046104 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157578 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046084 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046058 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560740 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157597 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560752 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560737 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157534 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157524 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157494 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157431 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046204 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560748 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560764 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560777 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560753 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157428 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560776 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157470 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157445 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560727 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157545 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560762 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157516 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157448 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157573 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157483 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046107 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157434 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046089 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560750 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560734 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157598 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560760 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046185 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157606 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157590 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560767 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157463 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046047 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P17046068 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10560765 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31157550 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03322001 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P20784905 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12800896 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12796257 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12799675 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03322001 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12799116 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12796657 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20784905 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P12799116 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P12796257 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1800006812.hg.1 | ENST00000580967.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006812.hg.1 | ENST00000580967.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006812.hg.1 | ENST00000584531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006812.hg.1 | ENST00000584531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006812.hg.1 | ENST00000584531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006812.hg.1 | ENST00000584531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006812.hg.1 | NR_104164.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006812.hg.1 | NR_104164.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006812.hg.1 | NR_104164.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006812.hg.1 | NR_104164.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006812.hg.1 | ENST00000580967.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006812.hg.1 | ENST00000580967.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006812.hg.1 | ENST00000584531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006812.hg.1 | ENST00000584531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006812.hg.1 | ENST00000584531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006812.hg.1 | ENST00000584531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006812.hg.1 | NR_104164.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006812.hg.1 | NR_104164.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006812.hg.1 | NR_104164.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800006812.hg.1 | NR_104164.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146676.hg.1 | ENST00000580967.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146676.hg.1 | ENST00000584531.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146676.hg.1 | NR_104164.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800146676.hg.1 | TC1800006812.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800146676.hg.1 | ENST00000580967.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146676.hg.1 | ENST00000584531.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800146676.hg.1 | NR_104164.1 | Refseq |
