Assay Details
Gene Symbol: GNAZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK126747.1 | - | 1 | 3453 | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | immunoglobulin lambda joining 3 | IGLJ3 | J3 | -172.8kb | Sense | Chr. 22 - 22904988 - 22905025 on NCBI Build GRCh38 |
Downstream |
RAB36, member RAS oncogene family |
RAB36 | 63.9kb | Sense | Chr. 22 - 23145298 - 23169285 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
