Assay Details
| Gene Symbol: LOC100130502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK126832.1 | - | 1 | 547 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calmodulin-lysine N-methyltransferase | CAMKMT |
C2orf34, CLNMT, CaM KMT, Cam, KMT |
-147.6kb | Anti-sense | Chr. 2 - 44361714 - 44774388 on NCBI Build GRCh38 |
| Downstream | SIX3 antisense RNA 1 | SIX3-AS1 | 17.7kb | Sense | Chr. 2 - 44940154 - 44941878 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144086 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224553 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144149 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144131 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144103 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15238889 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15238896 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224500 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144108 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144165 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144120 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144075 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144121 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144088 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224559 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224558 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144078 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224540 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15238891 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224545 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224477 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224544 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224523 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144077 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224504 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15238898 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144137 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144162 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224459 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144141 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224556 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144147 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224522 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224555 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224577 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224548 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144068 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144072 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144126 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144125 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224471 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144142 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144098 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144107 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144085 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144091 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224510 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144113 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224564 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224497 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224517 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224554 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224546 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144118 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224490 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15238921 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224534 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144150 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224505 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15238899 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144089 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144092 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144159 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144099 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144119 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144116 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144073 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224547 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144094 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144071 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224530 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15238902 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224479 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144139 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144138 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144079 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224461 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224538 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144069 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144074 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15238895 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144084 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224515 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144093 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144115 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15238901 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144082 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01144129 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21224491 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 776473 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 12146 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3255929 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00363422 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P522314 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 244744_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000354756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000354756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000354756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000430847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000430847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000432125.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000432125.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000432125.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000432125.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000432125.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000437916.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000437916.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000437916.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000437916.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000437916.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000437916.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000437916.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000437916.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000444871.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000444871.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000444871.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000444871.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000444871.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000560399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012447.hg.1 | ENST00000560399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000354756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000354756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000354756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000430847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000430847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000432125.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000432125.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000432125.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000432125.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000432125.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000437916.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000437916.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000437916.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000437916.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000437916.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000437916.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000437916.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000437916.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000444871.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000444871.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000444871.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000444871.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000444871.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000560399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012447.hg.1 | ENST00000560399.1 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 244744_at | - | - |
| Human Genome U133 Plus 2.0 Array | 244744_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244744_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244744_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244744_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244744_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244744_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244744_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244744_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244744_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244744_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244744_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244744_at7 | - | - |
| Human Genome U95 Set | 69027_PM_at | - | - |
| Human Genome U95 Set | 69027_at | - | - |
| Human Genome U133 Set | 244744_at | - | - |
| Human Genome U133 Set | 244744_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3271000 | AK126832.1 | human_transcriptome |

LOC100130502
Genomic Map