Assay Details
| Gene Symbol: AP3S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011543136.2 | XP_011541438.2 | 6 | 4646 | 95 | |
| XM_017009025.1 | XP_016864514.1 | 5 | 4574 | 95 | ||
| GenBank mRNA | AK126876.1 | - | 1 | 1456 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | autophagy related 12 | ATG12 |
APG12, APG12L, FBR93, HAPG12 |
-67.3kb | Anti-sense | Chr. 5 - 115828196 - 115841851 on NCBI Build GRCh38 |
| Downstream | laeverin | LVRN |
APQ, AQPEP, TAQPEP |
40.9kb | Sense | Chr. 5 - 115962440 - 116027602 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

AP3S1
Genomic Map