Assay Details
| Gene Symbol: DAAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127022.1 | - | 1 | 4350 | 70 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | DB305528.1 | - | 1 | Chr. 6 - 39822696 - 39823233 [-] on Build GRCh38 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kinesin family member 6 | KIF6 |
C6orf102, dJ1043E3.1, dJ137F1.4, dJ188D3.1 |
-93.3kb | Anti-sense | Chr. 6 - 39335100 - 39725405 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100505635 | LOC100505635 | 65.6kb | Anti-sense | Chr. 6 - 39888790 - 39897380 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

DAAM2
Genomic Map