Assay Details
Gene Symbol: LOC100131101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK127068.1 | - | 1 | 4450 | 83 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105373942 | LOC105373942 | -199.7kb | Anti-sense | Chr. 2 - 235746235 - 235791606 on NCBI Build GRCh38 | |
Downstream | translation initiation factor IF-2-like | LOC105373944 | 166.3kb | Anti-sense | Chr. 2 - 236161340 - 236164886 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
