Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT HG-U133+ PM Array Plate |
236973_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013466.hg.1 |
ENST00000448734.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013466.hg.1 |
ENST00000448734.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013466.hg.1 |
ENST00000448734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013466.hg.1 |
ENST00000448734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013466.hg.1 |
ENST00000448734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013466.hg.1 |
ENST00000448734.1 |
Ensembl |
Human Genome U133 Plus 2.0 Array |
236973_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
236973_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236973_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236973_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236973_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236973_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236973_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236973_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236973_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236973_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236973_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236973_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
236973_at5 |
- |
- |
Human Genome U95 Set |
74960_PM_at |
- |
- |
Human Genome U95 Set |
69791_PM_s_at |
- |
- |
Human Genome U95 Set |
74960_at |
- |
- |
Human Genome U95 Set |
69791_s_at |
- |
- |
Human Genome U133 Set |
236973_at |
- |
- |
Human Genome U133 Set |
236973_PM_at |
- |
- |