Assay Details
| Gene Symbol: LOC100128851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127423.1 | - | 1 | 2353 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | EPN2 antisense RNA 1 | EPN2-AS1 | -13.4kb | Anti-sense | Chr. 17 - 19296596 - 19306261 on NCBI Build GRCh38 | |
| Downstream | B9 domain containing 1 | B9D1 |
B9, EPPB9, MKS9, MKSR1 |
12.7kb | Anti-sense | Chr. 17 - 19335372 - 19378193 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100128851
Genomic Map