Assay Details
| Gene Symbol: LOC100127994 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127538.1 | - | 1 | 1955 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ATPase phospholipid transporting 9B (putative) | ATP9B |
ATPASEP, ATPIIB, HUSSY-20, NEO1L, hMMR1 |
-117.8kb | Sense | Chr. 18 - 79069275 - 79378391 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC284240 | LOC284240 | 76.3kb | Sense | Chr. 18 - 79573705 - 79591904 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100127994
Genomic Map