Assay Details
Gene Symbol: ST6GALNAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK127567.1 | - | 1 | 3049 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | small nucleolar RNA host gene 16 | SNHG16 | ncRAN | -12.2kb | Anti-sense | Chr. 17 - 76557764 - 76565348 on NCBI Build GRCh38 |
Downstream | ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1 | ST6GALNAC1 |
HSY11339, SIAT7A, ST6GalNAcI, STYI |
36.8kb | Sense | Chr. 17 - 76617769 - 76643838 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
