Assay Details
Gene Symbol: LOC400743 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK127830.1 | - | 2 - 3 | 354 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | peptidyl arginine deiminase 2 | PADI2 |
PAD-H19, PAD2, PDI2 |
-73.5kb | Anti-sense | Chr. 1 - 17066761 - 17119453 on NCBI Build GRCh38 |
Downstream | peptidyl arginine deiminase 1 | PADI1 |
HPAD10, PAD1, PDI, PDI1 |
11.1kb | Sense | Chr. 1 - 17205123 - 17246006 on NCBI Build GRCh38 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK127830.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15032868 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065285 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065309 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15032866 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065330 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15032870 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065283 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065291 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065284 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065275 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065325 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065342 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065274 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065287 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20082007 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20082047 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20082008 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065326 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065316 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20082026 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065337 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065334 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065322 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065312 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065314 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065318 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15032871 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20082043 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065298 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20082024 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065304 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065340 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065305 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065339 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20082038 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065281 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065302 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065301 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065292 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065315 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20082003 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065294 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065341 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065331 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065293 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065303 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065296 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065282 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065295 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20082014 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065329 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20082022 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065276 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00065320 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3275500 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 34332 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 25278 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3275495 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3275500 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00021149 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00021149 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P30056322 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100007101.hg.1 | ENST00000412427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007101.hg.1 | ENST00000412427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007101.hg.1 | ENST00000412427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007101.hg.1 | ENST00000412427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007101.hg.1 | ENST00000412427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007101.hg.1 | ENST00000412427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007101.hg.1 | ENST00000412427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007101.hg.1 | ENST00000412427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007101.hg.1 | ENST00000412427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007101.hg.1 | ENST00000412427.1 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1894911 | AK127830.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3294196 | AK127830.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1894911 | AK127830.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1894911 | AK127830.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 4860494 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_100232 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1894911 | AK127830.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1894911 | AK127830.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1894911 | AK127830.1 | human_transcriptome |
