Assay Details
| Gene Symbol: YARS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127879.1 | - | 1 | 1751 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | KIAA1522 | KIAA1522 | -39.4kb | Anti-sense | Chr. 1 - 32741839 - 32774970 on NCBI Build GRCh38 | |
| Downstream | S100P binding protein | S100PBP | S100PBPR | 86.0 | Anti-sense | Chr. 1 - 32816767 - 32858879 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

YARS
Genomic Map