Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012713.hg.1 |
ENST00000411708.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012713.hg.1 |
ENST00000411708.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012713.hg.1 |
ENST00000411708.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012713.hg.1 |
ENST00000411708.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012713.hg.1 |
ENST00000457311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012713.hg.1 |
ENST00000457311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012713.hg.1 |
ENST00000457311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012713.hg.1 |
ENST00000618051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012713.hg.1 |
ENST00000618051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012713.hg.1 |
ENST00000618051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012713.hg.1 |
ENST00000411708.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012713.hg.1 |
ENST00000411708.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012713.hg.1 |
ENST00000411708.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012713.hg.1 |
ENST00000411708.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012713.hg.1 |
ENST00000457311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012713.hg.1 |
ENST00000457311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012713.hg.1 |
ENST00000457311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012713.hg.1 |
ENST00000618051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012713.hg.1 |
ENST00000618051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012713.hg.1 |
ENST00000618051.1 |
Ensembl |
Human Genome U95 Set |
61551_at |
- |
- |
Human Genome U95 Set |
61551_PM_at |
- |
- |