Assay Details
| Gene Symbol: LOC646484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK128168.1 | - | 1 | 2746 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MGAT4 family member D | MGAT4D | GnT1IP | Anti-sense | Chr. 4 - 140442257 - 140499829 on NCBI Build GRCh38 | |
| Downstream | ELMO domain containing 2 | ELMOD2 | 9830169G11Rik | 22.8kb | Sense | Chr. 4 - 140524146 - 140553770 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15620193 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640583 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640589 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640616 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640599 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640621 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640592 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03479788 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03479819 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03479814 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640587 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640625 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03479787 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03479827 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640593 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03479812 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03479794 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03479818 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03479801 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640582 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640611 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640586 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640597 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03479831 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640603 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03479825 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640594 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640590 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03479786 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03479790 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640623 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03479784 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640588 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640617 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03479833 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15620198 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03479828 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640606 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640622 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23640596 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03479795 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03479820 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16881874 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01092245 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 244649_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008840.hg.1 | ENST00000624981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008840.hg.1 | ENST00000624981.1 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 244649_at | - | - |
| Human Genome U133 Plus 2.0 Array | 244649_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244649_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244649_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244649_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244649_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244649_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244649_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244649_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244649_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244649_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244649_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244649_at3 | - | - |
| Human Genome U95 Set | 79669_at | - | - |
| Human Genome U95 Set | 79669_PM_at | - | - |
| Human Genome U133 Set | 244649_PM_at | - | - |
| Human Genome U133 Set | 244649_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1789334 | AK128168.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1789334 | AK128168.1 | human_transcriptome |

LOC646484
Genomic Map