Assay Details
Gene Symbol: LINC00114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027065.1 | 4 - 5 | 259 | 65 | ||
NR_027066.1 | 3 - 4 | 296 | 65 | |||
GenBank mRNA | AY204744.1 | - | 2 - 3 | 84 | 65 | |
AY204745.1 | - | 3 - 4 | 296 | 65 | ||
AY204746.1 | - | 4 - 5 | 259 | 65 | ||
AY204750.1 | - | 1 - 2 | 1193 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
ERG, ETS transcription factor |
ERG |
erg-3, p55 |
-79.1kb | Sense | Chr. 21 - 38367261 - 38662045 on NCBI Build GRCh38 |
Downstream |
ETS proto-oncogene 2, transcription factor |
ETS2 | ETS2IT1 | 62.8kb | Anti-sense | Chr. 21 - 38805307 - 38824955 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC21000451.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR21005306.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21005306.hg.1 | TC21000451.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR21005302.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21005302.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21005302.hg.1 | TC21000451.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR21005305.hg.1 | ENST00000442549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21005305.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21005305.hg.1 | TC21000451.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | NR_027067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | NR_027067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | NR_027067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | NR_027067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008149.hg.1 | NR_027067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21005296.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21005296.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21005296.hg.1 | ENST00000442549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21005296.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21005296.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21005296.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21005296.hg.1 | TC21000451.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR21005295.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21005295.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21005295.hg.1 | ENST00000442549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21005295.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21005295.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21005295.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21005295.hg.1 | TC21000451.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR21005300.hg.1 | ENST00000426607.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21005300.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21005300.hg.1 | ENST00000442549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21005300.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21005300.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21005300.hg.1 | NR_027067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21005300.hg.1 | TC21000451.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2100154920.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154920.hg.1 | TC2100008149.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100154920.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154916.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154916.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154916.hg.1 | TC2100008149.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100154916.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154916.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154919.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154919.hg.1 | TC2100008149.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100154919.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154910.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154910.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154910.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154910.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154910.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154910.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154910.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154910.hg.1 | TC2100008149.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100154910.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154910.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154910.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154910.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154910.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154910.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154910.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154914.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154914.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154914.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154914.hg.1 | NR_027067.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154914.hg.1 | TC2100008149.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100154914.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154914.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154914.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154914.hg.1 | NR_027067.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154909.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154909.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154909.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154909.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154909.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154909.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154909.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154909.hg.1 | TC2100008149.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100154909.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154909.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154909.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154909.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154909.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100154909.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100154909.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000411989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000429621.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | ENST00000448579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | NR_027065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | NR_027066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | NR_027067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | NR_027067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | NR_027067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | NR_027067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008149.hg.1 | NR_027067.1 | Refseq |
