Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005294.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005294.hg.1 |
ENST00000426607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005294.hg.1 |
ENST00000429621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005294.hg.1 |
ENST00000442549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005294.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005294.hg.1 |
NR_027065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005294.hg.1 |
NR_027066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005294.hg.1 |
NR_027067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005294.hg.1 |
TC21000451.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000429621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000429621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000429621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000429621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000429621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000429621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
NR_027065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
NR_027065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
NR_027065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
NR_027065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
NR_027065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
NR_027065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
NR_027065.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
NR_027066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
NR_027066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
NR_027066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
NR_027066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
NR_027066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
NR_027066.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
NR_027067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
NR_027067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
NR_027067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
NR_027067.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008149.hg.1 |
NR_027067.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000429621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000429621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000429621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000429621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000429621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000429621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
NR_027065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
NR_027065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
NR_027065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
NR_027065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
NR_027065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
NR_027065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
NR_027065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
NR_027066.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
NR_027066.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
NR_027066.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
NR_027066.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
NR_027066.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
NR_027066.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
NR_027067.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
NR_027067.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
NR_027067.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
NR_027067.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008149.hg.1 |
NR_027067.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154908.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154908.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154908.hg.1 |
ENST00000429621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154908.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154908.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154908.hg.1 |
NR_027065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154908.hg.1 |
NR_027066.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154908.hg.1 |
NR_027067.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154908.hg.1 |
TC2100008149.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100154908.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154908.hg.1 |
ENST00000411989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154908.hg.1 |
ENST00000429621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154908.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154908.hg.1 |
ENST00000448579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154908.hg.1 |
NR_027065.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154908.hg.1 |
NR_027066.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154908.hg.1 |
NR_027067.1 |
Refseq |