Assay Details
Gene Symbol: LOC100131131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY358263.1 | - | 1 | 43 | 60 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | interleukin 1 receptor type 2 | IL1R2 |
CD121b, CDw121b, IL-1R-2, IL-1RT-2, IL-1RT2, IL1R2c, IL1RB |
-92.5kb | Sense | Chr. 2 - 101991805 - 102028544 on NCBI Build GRCh38 |
Downstream | interleukin 1 receptor like 2 | IL1RL2 |
IL-1Rrp2, IL-36R, IL1R-rp2, IL1RRP2 |
63.6kb | Sense | Chr. 2 - 102186973 - 102242906 on NCBI Build GRCh38 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY358263.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01366015 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01365940 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469497 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469514 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01365970 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01365941 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01365978 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15293208 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01366024 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01366012 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01366021 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01365957 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469460 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01366007 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469522 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469479 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469490 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469462 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469484 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01366022 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01365993 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469507 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469473 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01365962 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469496 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01366008 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01366020 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469481 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01366023 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469510 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469480 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01366026 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01365971 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469470 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01365995 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01366006 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469464 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01365964 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469468 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469493 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469506 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01365948 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01365974 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01366019 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469476 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01365987 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469503 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01365982 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01366011 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01365979 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469486 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469511 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469499 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469509 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01365966 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15293213 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469456 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469458 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01365999 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21469494 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P932536 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 40520 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 24888 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3310548 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P932536 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P932536 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8043994 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8043993 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3200261 | AY358263.1 | human_transcriptome |
