Assay Details
| Gene Symbol: UCHL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB209038.1 | - | 1 | 51 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | UCHL1 antisense RNA 1 (head to head) | UCHL1-AS1 | -261.0 | Anti-sense | Chr. 4 - 41220074 - 41256727 on NCBI Build GRCh38 | |
| Downstream | LIM and calponin homology domains 1 | LIMCH1 |
LIMCH1A, LMO7B |
97.0kb | Sense | Chr. 4 - 41359607 - 41700044 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

UCHL1
Genomic Map