| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000590802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000590802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000590802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000628453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000628453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000628453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
NM_001195790.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
NM_001195790.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
NM_001195790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000590802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000590802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000590802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000628453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000628453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000628453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
NM_001195790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
NM_001195790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
NM_001195790.1 |
Refseq |