Assay Details
Gene Symbol: SKCG-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY662656.1 | - | 1 | 1555 | 105 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | DEAD-box helicase 10 | DDX10 | HRH-J8 | -16.8kb | Sense | Chr. 11 - 108665025 - 108940930 on NCBI Build GRCh38 |
Downstream | chromosome 11 open reading frame 87 | C11orf87 |
LOH11CR1A, NEURIM1 |
461.9kb | Sense | Chr. 11 - 109422120 - 109433003 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28444994 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28444987 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018652 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018622 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018660 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018612 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28444971 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018642 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018615 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018620 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28445003 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018628 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018638 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018625 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018656 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018617 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018653 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018649 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28444993 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28444968 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018605 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28445004 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28444975 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018629 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018636 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018668 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28445010 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28445028 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28444965 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16500246 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018664 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28444997 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28444962 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018662 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28444995 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28445013 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018618 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018601 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018606 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28445007 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28445016 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018608 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018639 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28444976 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28445008 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28445006 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018609 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018631 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28445023 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28444972 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28444985 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08018630 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28445019 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 126793 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 60020 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3302756 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P19365178 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173352 | AB208789.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173352 | AK308105.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173352 | AK312856.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173352 | BC005911.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173352 | CR594806.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173352 | CR595480.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173352 | CR612478.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173352 | CR618704.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173352 | CR620432.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173352 | M55421.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173352 | M75883.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173352 | M75884.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173352 | NM_001007099.1 | refseq_mrna_human_NM |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173352 | NM_001007100.1 | refseq_mrna_human_NM |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173352 | NM_001007250.1 | refseq_mrna_human_NM |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173352 | NM_002979.3 | refseq_mrna_human_NM |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P19008653 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19365178 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7943708 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7943707 | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1100008992.hg.1 | ENST00000366160.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008992.hg.1 | ENST00000615020.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008992.hg.1 | ENST00000366160.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008992.hg.1 | ENST00000615020.1 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3311170 | - | - |
HumanHT-12 v4 Expression BeadChip | ILMN_3307972 | AY662656.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3311180 | AY662656.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1840354 | - | - |
Illumina Sentrix Human-6 v2 Expression BeadChip | 5340132 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_110527 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1840354 | - | - |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1840354 | - | - |
Illumina HumanWG-6_V2_0_R2 | ILMN_1840354 | - | - |
