Assay Details
| Gene Symbol: ZRANB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123114.1 | - | 1 | 176 | 89 | |
| CR749325.1 | - | 1 | 3249 | 89 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ZRANB2 antisense RNA 1 | ZRANB2-AS1 | Anti-sense | Chr. 1 - 71046506 - 71067184 on NCBI Build GRCh38 | ||
| Downstream | ZRANB2 antisense RNA 2 (head to head) | ZRANB2-AS2 | 12.6kb | Anti-sense | Chr. 1 - 71081324 - 71237723 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ZRANB2
Genomic Map