Assay Details
| Gene Symbol: LINC01181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK001351.1 | - | 1 - 2 | 180 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | BAALC antisense RNA 2 | BAALC-AS2 |
BAALCOS, C8orf56 |
Anti-sense | Chr. 8 - 103132963 - 103141342 on NCBI Build GRCh38 | |
| Downstream |
brain and acute leukemia, cytoplasmic |
BAALC | 7.3kb | Sense | Chr. 8 - 103140693 - 103230305 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC01181
Genomic Map