Assay Details
Gene Symbol: LINC01181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK001351.1 | - | 2 - 3 | 946 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | BAALC antisense RNA 2 | BAALC-AS2 |
BAALCOS, C8orf56 |
Anti-sense | Chr. 8 - 103132963 - 103141342 on NCBI Build GRCh38 | |
Downstream |
brain and acute leukemia, cytoplasmic |
BAALC | 5.4kb | Sense | Chr. 8 - 103140693 - 103230305 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
