Assay Details
| Gene Symbol: LINC01181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK001351.1 | - | 5 - 6 | 2191 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | BAALC antisense RNA 2 | BAALC-AS2 |
BAALCOS, C8orf56 |
Anti-sense | Chr. 8 - 103132963 - 103141342 on NCBI Build GRCh38 | |
| Downstream |
brain and acute leukemia, cytoplasmic |
BAALC | 338.0 | Sense | Chr. 8 - 103140693 - 103230305 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347057 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347104 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347126 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630197 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630242 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630218 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630245 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630207 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630178 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16122274 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347076 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630201 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347064 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630196 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630219 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630211 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630192 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630223 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347109 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630229 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16122277 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630191 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347100 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347112 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630206 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347119 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630199 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630193 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630253 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347066 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630220 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347059 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630177 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347061 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630204 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630222 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347063 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347085 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630238 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347054 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347106 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630188 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347074 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347110 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347092 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630217 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630189 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347080 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347098 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630246 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630210 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630228 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347083 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347107 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347121 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347118 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347124 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347115 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26630249 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347086 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06347105 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 259364 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 651860 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 973205 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 323727 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3362291 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P115434 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P115434 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 14093 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P16607299 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3362291 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P115434 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P115434 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P01984217 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 100006 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 13118 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 316 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18422459 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P16611350 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01984200 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01984217 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01984204 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18422436 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P16607299 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P115434 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P772798 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P01984204 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1562433_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 232326_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 232326_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1562433_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1562433_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562433_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232326_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232326_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562433_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562433_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562433_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562433_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562433_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562433_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232326_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232326_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232326_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232326_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562433_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232326_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232326_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562433_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232326_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562433_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232326_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232326_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562433_at4 | - | - |
| Human Genome U133 Set | 232326_PM_at | - | - |
| Human Genome U133 Set | 232326_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3273209 | AK001351.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3273209 | AK001351.1 | human_transcriptome |

LINC01181
Genomic Map