Assay Details
| Gene Symbol: MTHFD2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021544.1 | - | 1 | 1155 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | C-X-C motif chemokine ligand 2 | CXCL2 |
CINC-2a, GRO2, GROb, MGSA-b, MIP-2a, MIP2, MIP2A, SCYB2 |
-81.8kb | Anti-sense | Chr. 4 - 74097035 - 74099280 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105377276 | LOC105377276 | 95.1kb | Anti-sense | Chr. 4 - 74278045 - 74330322 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MTHFD2L
Genomic Map