Assay Details
| Gene Symbol: SRSF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021742.1 | - | 1 | 956 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat containing 40 | LRRC40 | dJ677H15.1 | -21.3kb | Anti-sense | Chr. 1 - 70144802 - 70205678 on NCBI Build GRCh38 |
| Downstream | ankyrin repeat domain 13C | ANKRD13C | dJ677H15.3 | 30.0kb | Anti-sense | Chr. 1 - 70259002 - 70354977 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SRSF11
Genomic Map