Assay Details
Gene Symbol: LOC101927770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110051.1 | 4 - 5 | 792 | 81 | ||
GenBank mRNA | AK024093.1 | - | 4 - 5 | 792 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007778.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000154007.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154007.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154007.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000154007.hg.1 | TC2000007778.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000154007.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154007.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154007.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000154012.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154012.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154012.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000154012.hg.1 | TC2000007778.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000154012.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154012.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154012.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000154005.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154005.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154005.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000154005.hg.1 | TC2000007778.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000154005.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154005.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154005.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000154006.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154006.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154006.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000154006.hg.1 | TC2000007778.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000154006.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154006.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154006.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000154009.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154009.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154009.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000154009.hg.1 | TC2000007778.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000154009.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154009.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154009.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000154004.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154004.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154004.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000154004.hg.1 | TC2000007778.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000154004.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154004.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154004.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000154010.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154010.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154010.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000154010.hg.1 | TC2000007778.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000154010.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154010.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154010.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007778.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000154011.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154011.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154011.hg.1 | NR_110051.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000154011.hg.1 | TC2000007778.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000154011.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154011.hg.1 | ENST00000424252.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000154011.hg.1 | NR_110051.1 | Refseq |
