Assay Details
| Gene Symbol: ABLIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK054890.1 | - | 1 | 1251 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | actin filament associated protein 1 like 2 | AFAP1L2 |
CTB-1144G6.4, KIAA1914, XB130 |
-19.3kb | Sense | Chr. 10 - 114280730 - 114405143 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ABLIM1
Genomic Map