Assay Details
Gene Symbol: LOC145945 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL137758.1 | - | 1 | 5254 | 90 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nuclear receptor subfamily 2 group F member 2 | NR2F2 |
ARP1, CHTD4, COUPTFB, COUPTFII, NF-E3, NR2F1, SVP40, TFCOUP2 |
-421.3kb | Anti-sense | Chr. 15 - 96325928 - 96340263 on NCBI Build GRCh38 |
Downstream | SPATA8 antisense RNA 1 (head to head) | SPATA8-AS1 | 5.1kb | Sense | Chr. 15 - 96772005 - 96783337 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360684 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360728 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856395 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856479 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16830320 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360676 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856396 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360762 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16830309 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856404 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360677 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360675 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360695 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360778 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360666 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360689 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16830331 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16830324 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360711 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856414 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360772 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16830326 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856451 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16830307 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360650 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360753 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856446 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856505 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360696 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16830310 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856447 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360740 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856483 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856457 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360739 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360729 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856477 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360669 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856500 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360750 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360679 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856470 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856386 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856497 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360690 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360727 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856430 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360707 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360698 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16830329 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360651 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856385 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856473 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360773 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360705 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360761 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360775 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856461 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360756 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856468 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360649 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360693 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856439 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856411 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856490 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856394 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856466 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360702 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856435 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360704 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360770 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856452 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16830332 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856405 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360667 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360661 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16830334 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360710 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856471 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856487 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360678 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360765 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360732 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856460 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856459 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360734 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856440 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856408 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856499 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16830323 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360724 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856489 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856423 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360688 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856384 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856388 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360682 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360653 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360699 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856494 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16830325 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856495 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360706 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856438 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360692 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856465 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856393 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360674 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856445 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360749 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09856482 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30360736 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16830311 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 513534 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3841755 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 20770 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3841755 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P20360636 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P40515031 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20360645 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20360636 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P701129 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P20360636 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 232577_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 232577_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 232577_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232577_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232577_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232577_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232577_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232577_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232577_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232577_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232577_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232577_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232577_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232577_at5 | - | - |
Human Genome U95 Set | 75721_PM_at | - | - |
Human Genome U95 Set | 75721_at | - | - |
Human Genome U133 Set | 232577_at | - | - |
Human Genome U133 Set | 232577_PM_at | - | - |
