Assay Details
Gene Symbol: NEURL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285486.1 | NP_001272415.1 | 5 - 6 | 953 | 86 | |
GenBank mRNA | AL389981.1 | - | 5 - 6 | 913 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | non-SMC condensin I complex subunit H | NCAPH |
BRRN1, CAP-H |
-120.6kb | Anti-sense | Chr. 2 - 96335741 - 96377095 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC105373496 | LOC105373496 | 28.7kb | Sense | Chr. 2 - 96527940 - 96532306 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NR_104346.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NR_104346.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NR_104346.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NR_104346.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NR_104346.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NR_104346.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013524.hg.1 | NR_104346.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200198947.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200198947.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200198947.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200198947.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200198947.hg.1 | TC0200013524.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200198942.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200198942.hg.1 | ENST00000435380.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200198942.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200198942.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200198942.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200198942.hg.1 | NR_104346.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200198942.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200198942.hg.1 | ENST00000435380.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200198942.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200198942.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200198942.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200198942.hg.1 | NR_104346.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200198942.hg.1 | TC0200013524.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000435380.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000444588.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NR_104346.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NR_104346.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NR_104346.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NR_104346.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NR_104346.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NR_104346.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013524.hg.1 | NR_104346.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200198944.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200198944.hg.1 | ENST00000435380.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200198944.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200198944.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200198944.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200198944.hg.1 | NR_104346.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200198944.hg.1 | ENST00000310865.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200198944.hg.1 | ENST00000435380.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200198944.hg.1 | ENST00000451794.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200198944.hg.1 | NM_001285485.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200198944.hg.1 | NM_001285486.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200198944.hg.1 | NR_104346.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200198944.hg.1 | TC0200013524.hg.1 | Clariom D TC |
