Assay Details
Gene Symbol: RAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK055170.1 | - | 1 | 1950 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MT-RNR2-like 3 | MTRNR2L3 | HN3 | -16.0kb | Anti-sense | Chr. 20 - 57358440 - 57359822 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC100291105 | LOC100291105 | 5.3kb | Anti-sense | Chr. 20 - 57383383 - 57393080 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
