Assay Details
Gene Symbol: LINC00942 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_028415.1 | 1 - 2 | 280 | 65 | ||
GenBank mRNA | AK090616.1 | - | 1 - 2 | 244 | 65 | |
BG332174.1 | - | 280 | 65 | |||
BQ942184.1 | - | 1 - 2 | 186 | 65 | ||
BQ954540.1 | - | 1 - 2 | 201 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ELKS/RAB6-interacting/CAST family member 1 | ERC1 |
Cast2, ELKS, ERC-1, RAB6IP2 |
-4.6kb | Sense | Chr. 12 - 990010 - 1495933 on NCBI Build GRCh38 |
Downstream | Wnt family member 5B | WNT5B | 26.4kb | Sense | Chr. 12 - 1617056 - 1647212 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000015.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
