Assay Details
| Gene Symbol: RSU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK092036.1 | - | 1 | 2150 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | complement C1q like 3 | C1QL3 |
C1QTNF13, C1ql, CTRP13, K100 |
-198.3kb | Sense | Chr. 10 - 16513743 - 16522005 on NCBI Build GRCh38 |
| Downstream | cubilin | CUBN |
IFCR, MGA1, gp280 |
101.1kb | Sense | Chr. 10 - 16823966 - 17130492 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RSU1
Genomic Map