Assay Details
Gene Symbol: FRMD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK093978.1 | - | 1 | 1549 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101928524 | LOC101928524 | -33.6kb | Anti-sense | Chr. 10 - 13650927 - 13655929 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC101928453 | LOC101928453 | 382.5kb | Anti-sense | Chr. 10 - 14074284 - 14087881 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
