Assay Details
| Gene Symbol: LOC100131864 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK094219.1 | - | 1 | 1251 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromodomain helicase DNA binding protein 5 | CHD5 | CHD-5 | Sense | Chr. 1 - 6101787 - 6180134 on NCBI Build GRCh38 | |
| Downstream | ribosomal protein L22 | RPL22 |
EAP, HBP15, HBP15/L22, L22 |
6.6kb | Sense | Chr. 1 - 6185020 - 6199619 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100131864
Genomic Map