Assay Details
| Gene Symbol: SCGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK094387.1 | - | 1 | 849 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | capping protein regulator and myosin 1 linker 1 | CARMIL1 |
CARMIL, CARMIL1a, LRRC16, LRRC16A, dJ501N12.1, dJ501N12.5 |
-79.8kb | Sense | Chr. 6 - 25279428 - 25620530 on NCBI Build GRCh38 |
| Downstream |
histone cluster 1, H2aa |
HIST1H2AA |
H2AA, H2AFR, TH2A, bA317E16.2 |
24.3kb | Anti-sense | Chr. 6 - 25726063 - 25726562 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SCGN
Genomic Map