Assay Details
| Gene Symbol: LOC285422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL832228.1 | - | 1 - 2 | 260 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SMAD family member 1 | SMAD1 |
BSP-1, BSP1, JV4-1, JV41, MADH1, MADR1 |
-26.9kb | Anti-sense | Chr. 4 - 145481306 - 145559176 on NCBI Build GRCh38 |
| Downstream | methylmalonic aciduria (cobalamin deficiency) cblA type | MMAA | cblA | 31.2kb | Anti-sense | Chr. 4 - 145619388 - 145660035 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500451 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662447 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500390 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662462 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500386 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500411 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662469 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662502 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662489 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662455 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500404 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500426 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500422 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662506 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500453 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662450 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500374 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662512 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662522 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500450 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500421 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500424 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662468 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500398 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662488 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500387 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662486 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662477 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500400 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662453 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662464 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662523 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500427 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500428 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662518 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662487 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662460 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662481 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662511 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662452 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662482 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500430 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662505 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662513 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662466 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662500 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500449 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662521 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500443 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500420 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662514 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500436 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23662454 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500402 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03500384 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 109393 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 112502 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3699565 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 58196 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3699565 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 33179 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16893157 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16893166 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1563685_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1563685_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1563685_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563685_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563685_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563685_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563685_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563685_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563685_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563685_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563685_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563685_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563685_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563685_at1 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1857510 | AL832228.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1857510 | AL832228.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1857510 | AL832228.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 2570349 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_88596 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1857510 | AL832228.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1857510 | AL832228.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1857510 | AL832228.1 | human_transcriptome |

LOC285422
Genomic Map