Assay Details
| Gene Symbol: SETDB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL833957.1 | - | 1 | 52 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calcium binding protein 39 like | CAB39L |
MO25-BETA, MO2L, bA103J18.3 |
-47.3kb | Anti-sense | Chr. 13 - 49308628 - 49444346 on NCBI Build GRCh38 |
| Downstream | PHD finger protein 11 | PHF11 |
APY, BCAP, IGEL, IGER, IGHER, NY-REN-34, NYREN34 |
3.5kb | Sense | Chr. 13 - 49495610 - 49528992 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SETDB2
Genomic Map