Assay Details
| Gene Symbol: USP46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021912.1 | - | 1 | 2845 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | spermatogenesis associated 18 | SPATA18 |
Mieap, SPETEX1 |
-490.9kb | Anti-sense | Chr. 4 - 52051331 - 52097305 on NCBI Build GRCh38 |
| Downstream | USP46 antisense RNA 1 | USP46-AS1 | 68.3kb | Anti-sense | Chr. 4 - 52659406 - 52661498 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

USP46
Genomic Map