Assay Details
Gene Symbol: MAPK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK022161.1 | - | 1 | 850 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Rho GTPase activating protein 24 | ARHGAP24 |
FILGAP, RC-GAP72, RCGAP72, p73, p73RhoGAP |
-61.2kb | Sense | Chr. 4 - 85475131 - 86002670 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC101929064 | LOC101929064 | 54.8kb | Sense | Chr. 4 - 86119806 - 86219926 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
