Assay Details
Gene Symbol: LOC100129312 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK095037.1 | - | 1 | 1950 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Sp3 transcription factor | SP3 | SPR2 | -21.9kb | Sense | Chr. 2 - 173906459 - 173965702 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC100128905 | LOC100128905 | 34.5kb | Anti-sense | Chr. 2 - 174025280 - 174027158 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684948 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684859 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684916 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808956 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684940 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808914 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808908 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684912 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684958 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684851 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808970 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808979 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808961 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684865 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684878 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684942 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684932 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684889 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684876 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684917 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684970 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808913 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808947 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684891 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808858 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684927 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684936 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684895 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684904 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684967 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684964 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684877 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808894 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808937 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808946 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808864 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808965 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684881 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808963 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15356886 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684971 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684935 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808955 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808876 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684900 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808944 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808969 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684905 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808875 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684899 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808892 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808904 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684846 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808871 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684924 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684926 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684843 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684969 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684966 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808951 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684879 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684946 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808987 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808907 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684953 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684963 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684980 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684984 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684862 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808891 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684913 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684863 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684910 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684952 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684921 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808909 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684915 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808896 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684902 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684933 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684853 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808921 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684959 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684908 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808860 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808964 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684845 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808912 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808893 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684861 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808942 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684883 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808966 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684918 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684923 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808945 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684850 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808916 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808952 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684855 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684977 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808971 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808976 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684975 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684885 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15356887 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21808895 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01684976 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 831910 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P349728 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P349728 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P384823 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 2079 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 28725 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 24051 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_24_P349728 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P384823 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P349728 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P349728 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P384823 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P30772486 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P900879 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | XM_001725750_at | - | - |
Affymetrix Human Gene 1.0 ST Array | XM_001721824_at | - | - |
Affymetrix Human Gene 1.0 ST Array | XM_001721839_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8056811 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8056810 | - | - |
